Learn More
Memmert™ Full-Sight Glass Door for UN260plus, Max. 300°C
Full-sight glass door constructed of four insulated glass layers.
Brand: Memmert™ UN260plusB1
This item is not returnable.
View return policy
Description
- Accessory
- Full-sight
- Glass (4 layers)
- Temperature range up to 300°C
Specifications
UN260plus Oven | |
300°C (Max.) | |
Full-sight Door | |
Glass |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Memmert™ Full-Sight Glass Door for UN260plus, Max. 300°C > For Use With: UN260plus
Spot an opportunity for improvement?Share a Content Correction