Learn More
Fulvestrant, MedChemExpress
Product Code. 16406266
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
Product Code. 16406266
Brand: Medchem Express HY13636
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress Fulvestrant (ICI 182780) is a pure antiestrogen and a potent estrogen receptor (ER) antagonist with an IC50 of 9.4 nM. Fulvestrant is also a GPR30 agonist. Fulvestrant effectively inhibits the growth of ER-positive MCF-7 cells with an IC50 of 0.29 nM. Fulvestrant also induces autophagy and has antitumor efficacy.
Fulvestrant (ICI 182780) is a pure antiestrogen and a potent estrogen receptor (ER) antagonist with an IC50 of 9.4 nM. Fulvestrant is also a GPR30 agonist. Fulvestrant effectively inhibits the growth of ER-positive MCF-7 cells with an IC50 of 0.29 nM. Fulvestrant also induces autophagy and has antitumor efficacy.
Specifications
Chemical Name or Material Fulvestrant
CAS 129453-61-8
Color White
Purity Grade Notes Research
Molecular Formula C32H47F5O3S
Quantity 100 mg
Synonym ICI 182780 ZD 9238 ZM 182780
Solubility Information DMSO : 250 mg/mL (412.02 mM; Need ultrasonic)
SMILES C[C@@]12[C@@H](O)CC[C@@]1([H])[C@]3([H])[C@H](CCCCCCCCCS(CCCC(F)(F)C(F)(F)F)=O)CC4=C(C=CC(O)=C4)[C@@]3([H])CC2
Molecular Weight (g/mol) 606.77
Formula Weight 606.77
Percent Purity 95.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death