Learn More
Trajan™ Sealing Ring for GC Ferrule
Graphite sealing ring for use with GC Ferrules
Brand:  Trajan™ 0726520
 View more versions of this product
Product Code. 10641983
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • For use with GC Ferrules
  • 100% Graphite Sealing ring for 1/4 in. fittings
  • Supplied in packs of 10
Specifications
Specifications
1/16 in.
1.7 mm
450°C
3.2 mm
1.59 mm
100% Graphite
Sealing Ring