Learn More
Trajan Scientific and Medical™ GC Tubing Fused Silica (Not Deactivated)
Helps in maintaining a high performance chromatography system
Brand: Trajan Scientific and Medical™ 062472
This item is not returnable.
View return policy
Description
- High quality fused silica tubing
- Tubing protected with high temperature polyimide resin
- Used in a wide range of GC, LC and biotechnology applications
Specifications
Tubing | |
0.15 mm | |
10 m |
GC, LC and Biotechnology | |
0.22 mm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Trajan Scientific and Medical™ GC Tubing Fused Silica (Not Deactivated) > O.D. x I.D. x L: 0.22 x 0.15mm x 10m
Spot an opportunity for improvement?Share a Content Correction