Learn More
Trajan Scientific and Medical™ GC Tubing Fused Silica (Not Deactivated)
Helps in maintaining a high performance chromatography system
Brand: Trajan Scientific and Medical™ 0624685
Description
- High quality fused silica tubing
- Tubing protected with high temperature polyimide resin
- Used in a wide range of GC, LC and biotechnology applications
Specifications
Tubing | |
0.1 mm | |
10 m |
GC, LC and Biotechnology | |
0.2 mm |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.