Learn More
Trajan Scientific and Medical™ GC Tubing Fused Silica (Not Deactivated)
Helps in maintaining a high performance chromatography system
Brand:  Trajan Scientific and Medical™ 0624685
 View more versions of this product
Product Code. 10645473
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
This item is not returnable. View return policy
Description
Description
  • High quality fused silica tubing
  • Tubing protected with high temperature polyimide resin
  • Used in a wide range of GC, LC and biotechnology applications
Specifications
Specifications
Tubing
0.1 mm
10 m
GC, LC and Biotechnology
0.2 mm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Trajan Scientific and Medical™ GC Tubing Fused Silica (Not Deactivated) > O.D. x I.D. x L: 0.2 x 0.1mm x 10m