Learn More
Trajan Scientific and Medical™ GC Tubing Fused Silica (Not Deactivated)
Product Code. 13184359
Click to view available options
Overall Length (Metric):
1 m
10 m
25 m
Diameter (Metric) Inner:
0.01 mm
0.025 mm
0.04 mm
0.05 mm
0.06 mm
0.075 mm
0.1 mm
0.11 mm
0.15 mm
0.22 mm
0.25 mm
0.32 mm
0.53 mm
Diameter (Metric) Outer:
0.14 mm
0.15 mm
0.17 mm
0.2 mm
0.22 mm
0.285 mm
0.363 mm
0.43 mm
0.68 mm
Unit Size:
10m
25m
Each
Product Code. 13184359
Brand: Trajan Scientific and Medical™ 062711
Only null left
Add to basket
Add to basket
Helps in maintaining a high performance chromatography system
  • High quality fused silica tubing
  • Tubing protected with high temperature polyimide resin
  • Used in a wide range of GC, LC and biotechnology applications
Specifications
Type Tubing
For Use With (Application) GC, LC and Biotechnology
Diameter (Metric) Inner 0.05 mm
Diameter (Metric) Outer 0.363 mm
Overall Length (Metric) 1 m