Learn More
Trajan Scientific and Medical™ GC Tubing Fused Silica (Not Deactivated)
Click to view available options
Overall Length (Metric):
1 m
10 m
25 m
Diameter (Metric) Inner:
0.01 mm
0.025 mm
0.04 mm
0.05 mm
0.06 mm
0.075 mm
0.1 mm
0.11 mm
0.15 mm
0.22 mm
0.25 mm
0.32 mm
0.53 mm
Diameter (Metric) Outer:
0.14 mm
0.15 mm
0.17 mm
0.2 mm
0.22 mm
0.285 mm
0.363 mm
0.43 mm
0.68 mm
Unit Size:
10m
25m
Each
Description
- High quality fused silica tubing
- Tubing protected with high temperature polyimide resin
- Used in a wide range of GC, LC and biotechnology applications
Specifications
Specifications
| Type | Tubing |
| For Use With (Application) | GC, LC and Biotechnology |
| Diameter (Metric) Inner | 0.05 mm |
| Diameter (Metric) Outer | 0.22 mm |
| Overall Length (Metric) | 10 m |