Learn More
Trajan Scientific and Medical™ GC Tubing Fused Silica (Not Deactivated)
Product Code. 15298823
Click to view available options
Overall Length (Metric):
1 m
10 m
25 m
Diameter (Metric) Inner:
0.01 mm
0.025 mm
0.04 mm
0.05 mm
0.06 mm
0.075 mm
0.1 mm
0.11 mm
0.15 mm
0.22 mm
0.25 mm
0.32 mm
0.53 mm
Diameter (Metric) Outer:
0.14 mm
0.15 mm
0.17 mm
0.2 mm
0.22 mm
0.285 mm
0.363 mm
0.43 mm
0.68 mm
Unit Size:
10m
25m
Each
Product Code. 15298823
Brand: Trajan Scientific and Medical™ 0624655
Please to purchase this item. Need a web account? Register with us today!
Helps in maintaining a high performance chromatography system
  • High quality fused silica tubing
  • Tubing protected with high temperature polyimide resin
  • Used in a wide range of GC, LC and biotechnology applications
Specifications
Type Tubing
For Use With (Application) GC, LC and Biotechnology
Diameter (Metric) Inner 0.06 mm
Diameter (Metric) Outer 0.22 mm
Overall Length (Metric) 10 m