Learn More
Vilber Lourmat™ Germicidal UV Lamp
Brand: Vilber Lourmat™ 3121 2301 1
This item is not returnable.
View return policy
Specifications
Germicidal UV Lamp | |
60 W | |
140 mm | |
100 mm | |
980 mm | |
1 Ea. |
230 V | |
254 nm | |
60 W, 230 V, 50 Hz | |
50 Hz | |
152 mW/cm2 |