missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GFRAL Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5112099
This item is not returnable.
View return policy
Description
GFRAL Polyclonal Antibody for Western Blot
Brainstem-restricted receptor for GDF15 which regulates food intake, energy expenditure and body weight in response to metabolic and toxin-induced stresses (PubMed:28953886, PubMed:28846097, PubMed:28846098, PubMed:28846099). Upon interaction with its ligand, GDF15, interacts with RET and induces cellular signaling through activation of MAPK- and AKT- signaling pathways. [UniProt].
Specifications
| GFRAL | |
| Polyclonal | |
| PBS with no preservative | |
| Q6UXV0 | |
| GFRAL | |
| A synthetic peptide corresponding to the N-terminus of the Human GFRAL. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Western Blot | |
| Unconjugated | |
| GFRAL | |
| AY457637; bA360D14.1; C6orf144; GDNF family receptor alpha like; GDNF family receptor alpha-like; GDNF receptor alpha-like; GFRAL; Gral; IVFI9356; RGD1565220; UNQ9356; UNQ9356/PRO34128 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 389400 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction