missing translation for 'onlineSavingsMsg'
Learn More

GGsTop, Tocris Bioscience™

Product Code. 30002011
Click to view available options
Quantity:
10 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Inhibitor of γ-glutamyl transpeptidase (GGT)

Selective, irreversible γ-glutamyl transpeptidase (GGT) inhibitor; displays no inhibitory effects on glutamine amidotransferases or asparagine synthetase. Suppresses oxidative stress by inhibiting GGT activation. Prevents ischemia/reperfusion-induced acute kidney injury in rats in vivo.

Specifications

CAS 926281-37-0
Molecular Formula C13H18NO7P
Quantity 10 mg
Synonym ggstop, methyl carboxymethylphenyl aminocarboxypropylphosphonate, 2-amino-4-3-carboxymethyl phenyl methyl phosphono butanoic acid, 2-amino-4-methyl 3-carboxymethyl phenyl phosphono butanoic acid, 2-amino-4-3-carboxymethyl phenoxy methoxy phosphoryl butanoic acid, 3-3-amino-3-carboxypropyl methoxyphosphinyl oxy benzeneacetic acid
InChI Key NTFPDEDRMYYPAC-UHFFFAOYSA-N
SMILES COP(=O)(CCC(C(=O)O)N)OC1=CC=CC(=C1)CC(=O)O
IUPAC Name 2-amino-4-[[3-(carboxymethyl)phenoxy]-methoxyphosphoryl]butanoic acid
Molecular Weight (g/mol) 331.261
PubChem CID 23582776
Formula Weight 331.26
Percent Purity >98%
Chemical Name or Material GGsTop
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.