Learn More
Gibco™ LV-MAX™ Lentiviral Production System
Product Code. 15829081
Click to view available options
Quantity:
1 Kit
Unit Size:
1 set
For Research Use Only. All usage must comply with product instructions.
Product Code. 15829081
Brand: Gibco™ A35684
Please to purchase this item. Need a web account? Register with us today!
For Research Use Only. All usage must comply with product instructions.
The LV-MAX Lentiviral Production System is the first optimized system that provides a scalable and high-yield lentiviral vector production.
The Gibco LV-MAX Lentiviral Production System is an optimized, scalable system designed to maximize lentiviral vector production. This system is specifically designed for high-density suspension cultures of HEK293-derived Viral Production Cells, which are adapted to a chemically defined, serum-free, and protein-free LV-MAX Production Medium. The kit also includes a high-efficiency, cationic, lipid-based transfection reagent, LV supplement, and LV enhancer, designed for the transient transfection of high-density suspension-adapted Viral Production Cells. All components work synergistically to generate superior and functional lentiviral particles under serum-free conditions compared to conventional polyethylenimine (PEI).
Key features of the LV-MAX Lentiviral Production System
  • High LV yields—achieves functional titers up to 1 x 108 TU/mL (unconcentrated)
  • High-density growth—supports growth of Viral Production Cells at densities greater than 1 x 107 cells/mL for maximum vector output
  • AOF formulation—chemically defined, serum-free, and animal-origin-free medium ensures consistent performance by eliminating variability from undefined components
  • Ready-to-use—no media supplementation required; simplifies workflows and provides reliable, lot-to-lot reproducibility in lentivirus production
  • Expression system compatible—fully compatible with the LV-MAX Lentiviral Production System for seamless integration into end-to-end lentivirus manufacturing processes
  • Scalable and reproducible transfection and production—validated from 96–well plates to multi‐liter bioreactors, enabling easy scale-up without sacrificing cell yields
  • Easy and robust culture and transfection protocols, seamlessly integrating into any transient production workflow
Kit components
  • LV-MAX Viral Production Cells (VPC)—HEK293-derived cells adapted to high-density suspension culture, providing a robust platform for lentiviral vector production
  • LV-MAX Production Medium—a chemically defined, serum-free, protein-free medium, specially developed for high-density growth and transfection of Gibco Viral Production Cells
  • LV-MAX Transfection Reagent—a high-efficiency, cationic, lipid-based reagent uniquely formulated for co-transfecting multiple plasmids into high-density Viral Production Cell cultures. This reagent ensures high transfection efficiency with minimal toxicity.
  • LV-MAX Supplement—an optimized, chemically defined, serum-free, protein-free, animal origin-free formulation designed to suppress cell growth during transfection. It enhances lentiviral vector production without compromising cell viability or interfering with the production process.
  • LV-MAX Enhancer—a proprietary, animal origin-free formulation that significantly boosts lentiviral vector production
Applications
  • Gene therapy—ideal for the production of lentiviral vectors for gene therapy research and development
  • Cell therapy—supports the production of lentiviral vectors for cell therapy applications, enabling a seamless transition from discovery to clinical production
  • Research and development—provides a reliable and scalable solution for lentiviral vector production in various research settings
Components are available in RUO or CTS
Offered as both a Research Use Only (RUO) version and a GMP-compliant Cell Therapy Systems (CTS) version to support translational and clinical-scale applications.
TRUSTED_SUSTAINABILITY
Specifications
Cell Line Viral Production Cell-Line
Form Liquid
Product Type Lentiviral Production System
Product Line LV-MAX
Quantity 1 Kit
Shipping Condition Approved for shipment at Room Temperature or on Wet or Dry Ice
Content And Storage • 1 L LV-MAX Production Medium. Store at 2–8°C, protect from light.
• 0.3 L LV-MAX Transfection Kit. Store at 2–8°C, protect from light.
• 2 vials of Viral Production Cells. Store in liquid nitrogen until ready to use. Do not store the cells at -80°C.
Delivery Type Lentiviral, Transfection
Culture Type Suspension Cell Culture
Expression System Transient