Learn More
Extech General-Purpose RTD Temperature Probe
Shop All Cole-Parmer Products
missing translation for 'orderingAttributeHoverText'
Temperature Range (Metric):
-40°C to +500°C
missing translation for 'unitSize'
Each
Description
- Compatible with Extech models SDL200, Oyster-10/15/16 meters, 407227/407228, and PH210 depending on the probe.
- Expand the capabilities of your temperature meter with additional probes for a variety of different applications.
Specifications
Specifications
| Type | Probe |
| For Use With (Application) | General-purpose |
| Temperature Range (English) | -40°F to +932°F |
| Temperature Range (Metric) | -40°C to +500°C |