Learn More
Hirschmann™ Class A Volumetric Flasks with Poly Stopper
Product Code. 11962043
Click to view available options
Capacity (Metric):
10 mL
10,000 mL
100 mL
1000 mL
20 mL
200 mL
2000 mL
25 mL
250 mL
5 mL
50 mL
500 mL
5000 mL
Standard Taper Joint:
10/19 NS Standard Ground
12/21 NS Standard Ground
14/23 NS Standard Ground
19/26 NS Standard Ground
19/32 NS Standard Ground
24/29 NS Standard Ground
29/32 NS Standard Ground
34/35 NS Standard Ground
45/40 NS Standard Ground
7/16 NS Standard Ground
NS 14/23
Unit Size:
Each
Pack of 2
Product Code. 11962043
Brand: Hirschmann™ 2822185
Please to purchase this item. Need a web account? Register with us today!
Achieve precise standard solutions. Hirschmann™ Class A Volumetric Flasks with Poly Stopper is standardized to contain or to deliver a definite amount of liquid. Flask has silver diffusion ink with brown graduations.
  • With polyethylene stopper
  • With amber stain graduation
  • Acc. to DIN EN ISO 1042
  • Corrosion-free and resistant to thermal shock
  • Conformity certified, with dated batch identification
Specifications
Certifications/Compliance DIN EN ISO 1042
Closure Material Polyethylene
Capacity (Metric) 200 mL
Material Borosilicate Glass, Duran Glass
Height (Metric) 210 mm
Limit of Error ±0.15 mL
Standard Taper Joint 14/23 NS Standard Ground
Tolerance ±0.15 mL
Graduated Yes
Class A