Learn More
Hecht Karl™ Glass Class AS Volumetric Pipet
Brand:  Hecht Karl™ 42556044
 View more versions of this product
Product Code. 10135721
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
This item is not returnable. View return policy
Description
Description
Specifications
Specifications
Yes
AS
Yes
1 Pack
8 mL
DIN EN ISO 648, Conformity certified
Glass
Amber
0.02 mL
Volumetric Pipet