Learn More
Hecht Karl™ Glass Gas Washing Bottles
Click to view available options
Capacity (Metric):
100 mL
250 mL
Unit Size:
Each
Specifications
Specifications
| Capacity (Metric) | 250 mL |
| Material | Glass |
| Type | Gas Washing Bottle |
| Includes | Filter Plate |
| Pore Size | 1 μm |
| Certifications/Compliance | DIN 12596 |
| Quantity | 1 Each |