Learn More
Hirschmann™ Glass Trapezoidal Volumetric Flask Class A with Glass Stopper
Product Code. p-8011847
Click to view available options
Capacity (Metric):
1 mL
2 mL
5 mL
10 mL
20 mL
25 mL
Standard Taper Joint:
NS 10/19
NS 7/16
Unit Size:
Pack of 2
Product Code. 13147959
Brand: Hirschmann™ 2960253
Only null left
Add to basket
Add to basket
For volumetric accuracy and precision in liquid handling.
  • Class A
  • Manufactured from DURAN™ borosilicate glass
  • Blue graduation marks
  • Certified conformity, with dated batch identification
  • Trapezoidal shape
  • Standard ground and glass stopper
  • Calibrated for IN
  • Acc. to DIN EN ISO 1042
Specifications
Certifications/Compliance DIN EN ISO 1042
Closure Material Glass
Capacity (Metric) 5 mL
Material Duran Glass
Height (Metric) 70 mm
Limit of Error ±0.04 mL
Standard Taper Joint NS 10/19
Tolerance ±0.04 mL
Graduated Yes
Class A