Learn More
Hirschmann™ Class A USP Volumetric Flasks
Product Code. 10067602
Click to view available options
Capacity (Metric):
5 mL
10 mL
20 mL
25 mL
50 mL
100 mL
200 mL
250 mL
500 mL
1000 mL
2000 mL
Unit Size:
Each
Pack of 2
Product Code. 10067602
Brand: Hirschmann™ 282019327
Only null left
Add to basket
Add to basket
Allows for precise, safe liquid measurement and handling in the laboratory. Hirschmann Instruments™ Class A USP Volumetric Flasks are manufactured from durable DURAN™ glass tubing and are available with a Batch Certificate of Performance, an Individual USP-Certificate of Performance, and a DAkkS-Certificate.
  • Blue graduation
  • With dated batch identification
  • With NS ground and polyethylene stopper
Specifications
Certifications/Compliance USP
Closure Material Polyethylene
Capacity (Metric) 1000 mL
Material Borosilicate Glass, Duran Glass
Height (Metric) 300 mm
Limit of Error ±0.3 mL
Tolerance ±0.3 mL
Graduated Yes
Class A