Learn More
Carl Roth™ Gloves
Product Code. 10533814
Encompass
Click to view available options
Size:
8
Small
Unit Size:
2 pairs
Pack of 100
This item is not returnable. View return policy
Product Code. 10533814
Brand: Carl Roth™ EP07.1
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy
Specifications
Product Type Gloves
Size 8
For Use With (Application) Light assembly work in the automotive, plastics, textile and consumer goods industries, warehousing and logistics operations. All components acc. FDA 21 CFR 177.2600. Washable to 40 ° C.
Quantity 2Each