Learn More
Glycidol, 96%, Thermo Scientific Chemicals
51.76 € - 658.28 €
Chemical Identifiers
CAS 556-52-5
Molecular Formula C3H6O2
Molecular Weight (g/mol) 74.08
MDL Number MFCD00005147,MFCD00074873
InChI Key CTKINSOISVBQLD-UHFFFAOYNA-N
Synonym glycidol, oxiranemethanol, 2,3-epoxy-1-propanol, epihydrin alcohol, glycide, oxiranylmethanol, 3-hydroxypropylene oxide, 1-propanol, 2,3-epoxy, 2,3-epoxypropanol, glycidyl alcohol
PubChem CID 11164
ChEBI CHEBI:30966
IUPAC Name oxiran-2-ylmethanol
SMILES OCC1CO1
View More Specs
Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10590931
View Documents
Thermo Scientific Acros
120051000
100 mL Glass bottle
51.76 €
100mL
Please to purchase this item. Need a web account? Register with us today!
10550771
View Documents
Thermo Scientific Acros
120055000
500 mL Glass bottle
174.87 €
500mL
Please to purchase this item. Need a web account? Register with us today!
11924761
View Documents
Thermo Scientific Acros
120050025
2.5 L Glass bottle
658.28 €
2.50 L
Please to purchase this item. Need a web account? Register with us today!
Description
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Chemical Identifiers
556-52-5
74.08
CTKINSOISVBQLD-UHFFFAOYNA-N
11164
oxiran-2-ylmethanol
C3H6O2
MFCD00005147,MFCD00074873
glycidol, oxiranemethanol, 2,3-epoxy-1-propanol, epihydrin alcohol, glycide, oxiranylmethanol, 3-hydroxypropylene oxide, 1-propanol, 2,3-epoxy, 2,3-epoxypropanol, glycidyl alcohol
CHEBI:30966
OCC1CO1
Specifications
556-52-5
Beige-Yellow to White
160.0°C to 161.0°C
Authentic
1.4300 to 1.4340
17,104
1.11
Solubility in water: soluble. Other solubilities: soluble in alcohol and ether
OCC1CO1
74.08
CHEBI:30966
74.08
Powder, Crystals or Chunks
-54.0°C
1.1100g/mL
71°C
C3H6O2
MFCD00005147,MFCD00074873
15,4525
glycidol, oxiranemethanol, 2,3-epoxy-1-propanol, epihydrin alcohol, glycide, oxiranylmethanol, 3-hydroxypropylene oxide, 1-propanol, 2,3-epoxy, 2,3-epoxypropanol, glycidyl alcohol
CTKINSOISVBQLD-UHFFFAOYNA-N
oxiran-2-ylmethanol
11164
4.0 mPa.s (20°C)
96%
Glycidol
Videos
Safety and Handling
Safety and Handling
GHS H Statement
Suspected of causing genetic defects.
May cause cancer.
Causes skin irritation.
May cause respiratory irritation.
Harmful in contact with skin.
Harmful if swallowed.
Causes serious eye irritation
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF INHALED:
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 209-128-3
Product Identifier
  • Glycidol
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Acute toxicity Category 4
  • Carcinogenicity Category 1B
  • Serious eye damage/eye irritation Category 2
  • Germ cell mutagenicity Category 2
  • Reproductive toxicity Category 1B
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity after repeated exposure Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H350-May cause cancer.
  • H360F-May damage fertility.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H330-Fatal if inhaled.
  • H341-Suspected of causing genetic defects.
  • H373-May cause damage to organs.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P362-Take off contaminated clothing.