Learn More
Thermo Scientific Chemicals Glycine, 0.2M buffer soln., pH 2.5
63.10 € - 104.00 €
Chemical Identifiers
CAS | 56-40-6 |
---|---|
Molecular Formula | C2H5NO2 |
Molecular Weight (g/mol) | 75.07 |
MDL Number | MFCD00008131 |
InChI Key | DHMQDGOQFOQNFH-UHFFFAOYSA-N |
IUPAC Name | 2-aminoacetic acid |
SMILES | NCC(O)=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15454349
|
Thermo Scientific Chemicals
J61855.AK |
250 mL |
63.10 €
250mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15464349
|
Thermo Scientific Chemicals
J61855.AP |
500 mL |
104.00 €
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This buffer solution is used for eluting (dissociating) antibodies in various antibody purification procedures such as affinity purification of antibody using peptide coupled to a column. It is widely used as a buffer for a variety of immunological applications. Glycine is also used as an eluent in column chromatography. Glycine has been reported for use in the enzymatic determination of lactate using lactic dehydrogenase.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsThis buffer solution is used for eluting (dissociating) antibodies in various antibody purification procedures such as affinity purification of antibody using peptide coupled to a column. It is widely used as a buffer for a variety of immunological applications. Glycine is also used as an eluent in column chromatography. Glycine has been reported for use in the enzymatic determination of lactate using lactic dehydrogenase.
Solubility
Soluble in water (100 mg/ml), and acetone (slightly soluble). Insoluble in alcohol, and ether.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Recommended storage temperature 2 - 8°C
Chemical Identifiers
56-40-6 | |
75.07 | |
DHMQDGOQFOQNFH-UHFFFAOYSA-N | |
NCC(O)=O |
C2H5NO2 | |
MFCD00008131 | |
2-aminoacetic acid |
Specifications
56-40-6 | |
2.5 | |
C2H5NO2 | |
MFCD00008131 | |
NCC(O)=O | |
75.07 |
0.2 M | |
Liquid | |
Odorless | |
DHMQDGOQFOQNFH-UHFFFAOYSA-N | |
2-aminoacetic acid | |
Glycine |
Safety and Handling
Recommended Storage : Ambient temperatures
RUO – Research Use Only