missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glycine Receptor alpha 3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-30930
This item is not returnable.
View return policy
Description
Glycine Receptor alpha 3 Polyclonal specifically detects Glycine Receptor alpha 3 in Human samples. It is validated for Western Blot.
Specifications
| Glycine Receptor alpha 3 | |
| Polyclonal | |
| Unconjugated | |
| 0.1M Tris-Glycine (pH7), 10% Glycerol with 0.01% Thimerosal | |
| GLRA3 | |
| Carrier-protein conjugated synthetic peptide encompassing a sequence within the N-terminus region of human Glycine Receptor alpha 3. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Tested in human. Not yet tested in any other species. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot | |
| 1.0mg/mL | |
| Western Blot 1:500-1:3000 | |
| glycine receptor alpha 3 subunit, glycine receptor subunit alpha-3, glycine receptor, alpha 3, glycine receptor, alpha-3 polypeptide, ligand gated ion channel | |
| Rabbit | |
| 54 kDa | |
| 100 μL | |
| Signal Transduction | |
| 8001 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction