missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Goat anti-Human IgE Fc Secondary Antibody, HRP conjugate
Goat anti-Human IgE Fc Secondary Antibody, HRP conjugate
Brand: Invitrogen H15707
This item is not returnable.
View return policy
Description
Human IgE Fc Secondary Antibody for Flow![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Flow Cytometry | |
0.6mg/mL | |
Goat anti-Human IgE Fc Secondary Antibody, 1mL, 0.6mg/mL Concentration, Liquid, Goat Host, Human Species Reactivity, HRP Conjugate, IgG Isotype, 4°C Storage Temperature, PBS with 4mg/mL BSA, 40% glycerol Storage Buffer, For Flow Cytometry | |
Goat | |
1 mL | |
Secondary | |
4°C | |
IgG |
Polyclonal | |
HRP | |
Assay-dependent | |
Purified | |
RUO | |
1 Each | |
Liquid | |
HRP, AP, fluorescence |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 11
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Goat anti-Human IgE Fc Secondary Antibody, HRP conjugate > 1mL
Analyte Specific Reagent. Analytical and performance characteristics are not established.
Spot an opportunity for improvement?Share a Content Correction