missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Goat anti-Rabbit IgG Fc Secondary Antibody, PE/Atto594, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NBP2-60661PEATT594
590.40 EUR valid until 2025-03-28
Use promo code "25306" to get your promotional price.
This item is not returnable.
View return policy
Description
IgG Fc Polyclonal antibody specifically detects IgG Fc in Rabbit samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Fluorophore-linked immunosorbent assay
Specifications
IgG Fc | |
Polyclonal | |
PBS | |
Goat anti-Rabbit IgG Fc fragment Secondary Antibody was produced by repeated immunization with rabbit IgG f(c) fragment in goat | |
500 μg | |
Adaptive Immunity, Immunology | |
Rabbit | |
Purified |
Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Fluorophore-Linked Immunosorbent Assay | |
PE/ATTO 594 | |
Goat | |
Immunogen affinity purified | |
RUO | |
Secondary | |
Store at 4°C in the dark. Do not freeze. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction