Learn More
GPR15 Mouse anti-Human, Clone: 367902, R&D Systems™
GREENER_CHOICE
Mouse Monoclonal Antibody has been used in 9 publications
Brand:  R&D Systems MAB3654-SP
Encompass
Product Code. 15716404
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
This item is not returnable. View return policy
Description
Description
GPR15 Monoclonal specifically detects GPR15 in Human samples. It is validated for Flow Cytometry, CyTOF-reported.