missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Grace™ Vydac™ C18 (218TP) HPLC Columns
Designed for the separation of peptides less than 4000 to 5000 MW and enzymatic digest fragments
4795.20 €
Specifications
Flow Rate | 5 mL/min |
---|---|
For Use With | LC/MS Separation |
Particle Size | 5 μm |
Pore Size | 300 Å |
Surface Area | 60 m²/g |
Description
- Provide reliable, high resolution of proteins and large peptides
- Relied upon in numerous commercial scale protein purifications
- Vydac TP is completely scalable to ton quantities
- Threaded
Specifications
5 mL/min | |
5 μm | |
60 m²/g | |
C18 | |
8% | |
Yes | |
Column |
LC/MS Separation | |
300 Å | |
L1 | |
Silica | |
Reversed Phase | |
Reversed Phase | |
Reverse Phase HPLC |