missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GRB2 Antibody (JE49-73), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-32403
This item is not returnable.
View return policy
Description
GRB2 Monoclonal antibody specifically detects GRB2 in Human,Monkey,Mouse,Rat samples. It is validated for Flow Cytometry,Western Blot,Immunocytochemistry/Immunofluorescence
Specifications
| GRB2 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:2000, Flow Cytometry 1:1000, Immunocytochemistry/Immunofluorescence 1:100 | |
| abundant SRC homology, Adapter protein GRB2, ASH, EGFRBP-GRB2, epidermal growth factor receptor-binding protein GRB2, Grb3-3, growth factor receptor-bound protein 2, growth factor receptor-bound protein 3, HT027, MST084, MSTP084, NCKAP2, Protein Ash, SH2/SH3 adapter GRB2 | |
| Recombinant protein within Human GRB2 aa 1-100/ 217. (Uniprot: P62993) | |
| 100 μL | |
| Angiogenesis, Breast Cancer, Cancer, Extracellular Matrix, Signal Transduction | |
| 2885 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Flow Cytometry, Western Blot, Immunocytochemistry/Immunofluorescence | |
| JE49-73 | |
| Unconjugated | |
| 1*TBS (pH7.4), 0.05% BSA and 40% Glycerol | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human, Monkey, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction