missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ GSK 2033

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
GSK 2033 is a potent LXR antagonist (pIC50 = 7.5). Enhances T-cell proliferation and blocks T 0901317 -antiproliferative activity on T-cells. Cell permeable.
Specifications
Specifications
| Chemical Name or Material | 2,4,6-Trimethyl-N-[[3′-(methylsulfonyl)[1,1′-biphenyl]-4-yl]methyl]-N-[[5-(trifluoromethyl)-2-furanyl]methyl]benzenesulfonamide |
| CAS | 1221277-90-2 |
| Quantity | 50 mg |
| Target | LXR-like Receptor Antagonists |
| Molecular Formula | C29H28F3NO5S |
| Formula Weight | 591.66 |
| Purity | >99% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction