missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GTF3C5 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11447-2-AP-150UL
This item is not returnable.
View return policy
Description
Lamina-associated polypeptide 2 (LAP2), isoform beta is a protein that in humans is encoded by the TMPO gene. LAP2 is an inner nuclear membrane (INM) protein. LAP2 plays a role in the regulation of nuclear architecture by binding lamin B1 and chromosomes. This interaction is regulated by phosphorylation during mitosis.Specifications
| GTF3C5 | |
| Polyclonal | |
| Unconjugated | |
| GTF3C5 | |
| GTF3C5, TF3C epsilon, TFIIIC 63 kDa subunit, TFiiiC2 63, TFIIIC63, TFIIICepsilon | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 362095, 70239, 9328 | |
| -20°C | |
| Liquid |
| Immunoprecipitation, Western Blot | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q8R2T8, Q9Y5Q8 | |
| Gtf3c5 | |
| GTF3C5 Fusion Protein Ag1999 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction