Learn More
Pacific Handy Cutter Guarded Spring Back Safety Cutter Left
Product Code. p-7241159
Click to view available options
Color:
Green
Red
Unit Size:
Pack of 12
Product Code. 17729037
Brand: Pacific Handy Cutter S4SL
Only null left
Add to basket
Add to basket
The S4S is a safety knife with automatic blade retraction and additional protective guard. Therefore, it is very safe.
  • The protective guard also serves as a guide, e.g. when topping cartons.
  • With robust integrated tape splitter and spare blade storage.
  • Tool-free blade change.
  • With practical eyelet for lanyard fastening or hanging.
  • The green model is suitable for right-handed users.
  • The red model is suitable for left-handed users.
Specifications
Product Type Safety Cutter
Disposable Reusable
Sterility Non-sterile
Color Red