Learn More
IKA™ H 66.53 Stainless Steel Temperature Sensor
Shop All IKA Products
Click to view available options
Diameter (Metric):
3 mm
Unit Size:
Each
Specifications
Specifications
| Diameter (Metric) | 3 mm |
| Model | H 66.53 |
| Temperature (Metric) | -10°C to +260°C |
| Type | Temperature Sensor |