Learn More
Macherey-Nagel™ Haloform Test Mixture in Methanol
Brand: Macherey-Nagel™ 722371
This item is not returnable.
View return policy
Specifications
For Headspace Analyses, for GC Environmental Analyses | |
Dichloromethane (795 μg/mL), Trichloromethane (75 μg/mL), 1,1,1-trichloroethane (67 μg/mL), Tetrachloromethane (80 μg/mL), Trichloroethene (73 μg/mL), Bromodichloromethane (100 μg/mL), Dibromochloromethane (122 μg/mL), Tetrachloroethene (81 μg/mL), Tribromomethane (145 μg/mL) | |
1 mL | |
Haloform Test Mixture in Methanol |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Macherey-Nagel™ Haloform Test Mixture in Methanol > Capacity: 1mL
Spot an opportunity for improvement?Share a Content Correction