Learn More
Horst™ HBST Model Heating Tape
Product Code. 10668462
Click to view available options
Length (Metric):
1 m
10 m
17 m
2 m
3 m
4 m
5 m
7 m
Unit Size:
Each
Product Code. 10668462
Brand: Horst™ 20709
Only null left
Add to basket
Add to basket
Specifications
Dimensions (L x W) 7 Lm
Dimensions (L x W x H) 7 Lm
Length (Metric) 7 m
Voltage 230 V
Electrical Requirements ∼230V Voltage
Model HBST
Wattage 350w
Width (Metric) 25mm
Product Type Heating Tape
Temperature (Metric) 250°C