Learn More
HDAC1 Monoclonal Antibody (OTI5F9), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA502193
Description
Histone deacetylase (HDAC) is responsible for the deacetylation of lysine residues on the N-terminal part of the core histones - H2A, H2B, H3, and H4. Histone deacetylation gives a tag for epigenetic repression and plays and important role in transcriptional regulation, cell cycle progression, and developmental events. Histone deacetylases act via the formation of large multiprotein complexes.Specifications
| HDAC1 | |
| Monoclonal | |
| Unconjugated | |
| HDAC1 | |
| GON-10, HD1, RPD3, RPD3L1 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 297893, 3065, 433759, 487309 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot | |
| OTI5F9 | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| O09106, Q13547, Q4QQW4 | |
| HDAC1 | |
| Full length human recombit protein of human HDAC1 produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Canine, Human, Mouse, Rat | |
| Antibody | |
| IgG2b |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.