Learn More
HDAC9 Mouse anti-Human, Clone: OTI7G9, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies CF805392
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
HDAC-9 is a histone deacetylase with an important role in the regulation of gene expression and hematopoiesis. A nuclear protein, HDAC-9 belongs to the histone deacetylase family with several isoforms and is inhibited by Trichostatin A (TSA) and suberoylanilide hydroxamic acid. It helps in deacetylation of lysine residues on the N-terminal part of the core histones (H2A, H2B, H3 and H4) and gives a tag for epigenetic repression and plays an important role in transcriptional regulation, cell cycle progression and developmental events. MEF2-dependent transcription is repressed by HDAC-9 by recruiting HDAC-1 and/or HDAC-3. It also inhibits skeletal myogenesis and is involved in heart development. Ubiquitous but variable expression is seen in brain and other tissues like heart, pancreas and also in human bladder carcinoma cells. Lack of HDAC-9 protein is found in a family with Peters anomaly, a central corneal leukoma.Specifications
| HDAC9 | |
| Monoclonal | |
| Unconjugated | |
| HDAC9 | |
| HD7, HD7b, HD9, HDAC, HDAC7, HDAC7B, HDAC9B, HDAC9FL, HDRP, MITR | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 9734 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Western Blot | |
| OTI7G9 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| Q9UKV0 | |
| HDAC9 | |
| Human recombit protein fragment corresponding to amino acids 181-460 of human HDAC9 produced in E.coli. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.