Learn More
Heidolph™ Hei-FLOW Core 120
This peristaltic pump is ideal for the simple conveying of all media.
Brand: Heidolph™ 523-50010-00
2181.34 EUR valid until 2025-04-13
Use promo code "24122" to get your promotional price.
Description
- The pump drive of this model makes it possible to switch between a single channel and multi-channel pump head. By simply flipping the switch, it can also be conveyed in an anti-clockwise direction.
- Depending on the adapted pump head, a flow rate of 0.83 to 861 mL/min for the single-channel and 0.005 to 364 mL/min for the multi-channel pump head is guaranteed. The Value 01 conveys with a control accuracy of ± 5% at a speed range of 10 to 120 rpm.
Specifications
EU Plug | |
LED | |
230 V |
Digital Variable Speed | |
IP33 |
Product Suggestions
Customers who viewed this item also viewed
Promotions
-
Multiple Manufacturers
Precision at Your Fingertips
Master liquid handling with ease.Promo Code: 24122Expires: 2025-04-13
Days
6Hours
13Minutes
52Seconds
10
Your input is important to us. Please complete this form to provide feedback related to the content on this product.