Learn More
Greiner Bio-One High Grade Polypropylene Pipet Tips
Product Code. 10020252
Click to view available options
Volume (Metric):
10 to 200 μL
100 to 1000 μL
Color:
Blue
Natural
Yellow
Quantity:
250/Pk.
4800/Pk.
500/Pk.
96/Pk.
Unit Size:
Pack of 250
Pack of 4800
Pack of 500
Pack of 960
Product Code. 10020252
Brand: Greiner Bio-One 739261
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Manufactured from high-grade polypropylene to give the optimum performance and fit. Greainer BioOne™ High Grade Polypropylene Pipet Tips are ideally suited for Gilson™ Pipetman models.
  • High breaking strength, dimensionally stable, heat-resistant up to approx. +140°C; cold-resistant down to -190°C
  • Autoclavable
  • Free of heavy metals
  • Conforms to the ISO 9001 standard
  • Tested for conformance with the international standard DIN ISO 8655
  • Liquid end is bevelled so less liquid remains on pipet tip, contact surface of pipetted liquids is reduced and pipetting accuracy increases
  • Ideally suited for Gilson™ Pipetman P20, P100, P200, F5, F10, F25, F50, F200, U200
TRUSTED_SUSTAINABILITY
Specifications
Volume (Metric) 10 to 200 μL
Color Yellow
Autoclavable Autoclavable
Certifications/Compliance ISO 9001, DIN ISO 8655
Format Bulk
Quantity 96/Pk.
For Use With (Equipment) Eppendorf, Gilson Pipetman P200, Gilson Pipetman P100, Gilson Pipetman P20
Material Polypropylene