missing translation for 'onlineSavingsMsg'
Learn More

Greiner Bio-One High Grade Polypropylene Pipet Tips

Product Code. 10702492
Click to view available options
Volume (Metric):
10 to 200 μL
100 to 1000 μL
Color:
Blue
Natural
Yellow
Quantity:
250/Pk.
4800/Pk.
500/Pk.
96/Pk.
Unit Size:
Pack of 250
Pack of 4800
Pack of 500
Pack of 960
This item is not returnable. View return policy

Product Code. 10702492

Brand: Greiner Bio-One 739280

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Manufactured from high-grade polypropylene to give the optimum performance and fit. Greainer BioOne™ High Grade Polypropylene Pipet Tips are ideally suited for Gilson™ Pipetman models.

  • High breaking strength, dimensionally stable, heat-resistant up to approx. +140°C; cold-resistant down to -190°C
  • Autoclavable
  • Free of heavy metals
  • Conforms to the ISO 9001 standard
  • Tested for conformance with the international standard DIN ISO 8655
  • Liquid end is bevelled so less liquid remains on pipet tip, contact surface of pipetted liquids is reduced and pipetting accuracy increases
  • Ideally suited for Gilson™ Pipetman P20, P100, P200, F5, F10, F25, F50, F200, U200
TRUSTED_SUSTAINABILITY

Specifications

Volume (Metric) 10 to 200 μL
Sterility Non-sterile
Color Yellow
Autoclavable Autoclavable
Certifications/Compliance ISO 9001, DIN ISO 8655
Format Rack
Quantity 4800/Pk.
For Use With (Equipment) Gilson Pipetman P100, Gilson Pipetman P20, Gilson Pipetman P200
Graduated No
Material Polypropylene
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.