Learn More
Greiner Bio-One High Profile Polystyrene Microplate Lids
GREENER_CHOICE
Product Code. p-7075458
Click to view available options
Sterility:
Non-sterile
Sterile
Condensation Rings:
Yes
No. per Pack:
1
No. per Case:
100
Unit Size:
Pack of 100
Product Code. 10701401
Brand: Greiner Bio-One 656171
Only null left
Add to basket
Add to basket
For use with non TC-treated 96-well microplates. Greiner Bio-one High Profile Polystyrene Microplate Lids is free of detectable DNase, RNase, human DNA.
  • Free of detectable DNase, RNase, human DNA
  • Non-Pyrogenic, non-cytotoxic
  • Offer protection against contamination and evaporation during sample storage and cell cultivation
TRUSTED_SUSTAINABILITY
Specifications
Color Clear
Material Polystyrene
Sterility Sterile
Height (Metric) 9 mm
Condensation Rings Yes
No. per Case 100
No. per Pack 1
For Research Use Only. Not for use in diagnostic procedures, unless otherwise indicated.