Learn More
Cytiva His Mag Sepharose™ Ni Magnetic Beads
Designed for efficient, small-scale purification/screening of histidine-tagged proteins from different sources Cytiva Lifescience™ His Mag Sepharose™ Ni Magnetic Beads enable high-capacity small-scale purification.
260.00 € - 2257.00 €
Specifications
Binding Property 50 mg Histidine-tagged protein/mL medium
View More Specs
Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11540894
View Documents
Cytiva
28-9673-90
5 x 1 mL
260.00 €
Pack of 5
In Stock
to see stock.
Add to basket
11545804
View Documents
Cytiva
28-9799-17
10 x 1 mL Dieser Artikel ist im Moment nicht verfügbar oder wird nicht mehr hergestellt.
Schauen Sie bitte auf unsere Produktseite nach möglichen Alternativen
N/A
17296876
View Documents
Cytiva
29104065
100 mL
2257.00 €
100mL
Estimated Shipment: 05-03-2025
to see stock.
Add to basket
Description
Description
Together with MagRack 6, a separation tool for handling the beads in microcentrifuge tubes, up to six samples can be processed in parallel. For higher throughput, larger numbers of samples can easily be screened in parallel on a robotic device.
  • High-capacity small-scale purification of histidine-tagged proteins from different sources
  • High purity and yield
  • Easy parallel screening of histidine-tagged proteins with high repeatability
  • Scalability dash (en) Simple capture of histidine-tagged proteins from small or large sample volumes (low microliter to milliliter scale)
Order Info
1mL of medium slurry is sufficient for five purification runs according to the recommended protocol
Specifications
Specifications
50 mg Histidine-tagged protein/mL medium
Videos
Safety and Handling
Safety and Handling
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3