Learn More
Hoefer™ PlateMate™ Washing System
Click to view available options
:
Washing system; Plate Mate
Description
- Two plate racks with handles and dust covers
- Polypropylene washing tank with lid
Order Info
Accessories are available separately (See Cat. No. 03-500-449 and -450).
Specifications
Specifications
| Device Holds | Four sizes of Glass or Alumina plates: 18 x 16cm, 10 x 12cm, 10 x 10.5cm or 10 x 8cm, Holds either 10, 18cm W plates or 20, 8cm W plates |
| Type | PlateMate™ Washing System |