Learn More
Hoefer™ PlateMate™ Washing System
Product Code. 11509633
Click to view available options
:
Washing system; Plate Mate
Product Code. 11509633
Brand: Hoefer™ SE100
Only null left
Add to basket
Add to basket
Complete plate washing system
  • Two plate racks with handles and dust covers
  • Polypropylene washing tank with lid
Order Info
Accessories are available separately (See Cat. No. 03-500-449 and -450).
Specifications
Device Holds Four sizes of Glass or Alumina plates: 18 x 16cm, 10 x 12cm, 10 x 10.5cm or 10 x 8cm, Holds either 10, 18cm W plates or 20, 8cm W plates
Type PlateMate™ Washing System