Learn More
Edmund Bühler GmbH™ Homogenizer Knife
Product Code. 10001021
Click to view available options
Capacity (Metric):
25 mL
50 mL
100 mL
150 mL
Unit Size:
Each
Product Code. 10001021
Brand: Edmund Bühler GmbH™ 0002 984
Please to purchase this item. Need a web account? Register with us today!
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Manufactured from stainless steel. Edmund Bühler GmbH™ Homogenizer Knife is for use with a variety of sample vessels.
  • Provides mechanical crushing of samples used with Homogenizer HO 4/A
  • Stainless steel construction
  • Spindle, knife and sample vessel must be matched in size.
Specifications
Product Type Homogenizer Knife
Capacity (Metric) 25 mL
Blade Material Stainless Steel