Learn More
Sperian™ Honeywell™ 2000 Comfort Series Respirators
Click to view available options
Model:
2110 FFP1
2110 FFP2
2211 FFP2V
Unit Size:
Pack of 20
Pack of 25
Description
- Folds flat packed in individually sealed bags for transportation and storage
- Dolomite tested, demonstrating the respirators' resistance to clogging over time
- With high efficiency exhalation valve for protection
- Double layer face seal provides comfort and protection
Specifications
Specifications
| Certifications/Compliance | EN 149 : 2001 A1: 2009 |
| Disposable | Single-use |
| Filter Type | FFP2 |
| Product Type | Respirator Protection |
| Size | Medium |
| With Exhalation Valve | Yes |
| For Use With (Application) | Agriculture, Automotive and Part Manufacturer, Building and Construction, Catering, Chemical Industries, Food Industries, Foundry, General Industry, Glass Industries, Green Spaces, Iron and Steel Industry Laboratory, Maintenance Medical and Pharmaceutical |
| Latex Free | Yes |
| Model | 2110 FFP2 |
| Quantity | 25/Pk. |
| Show More |