missing translation for 'onlineSavingsMsg'
Learn More
Sperian™ Honeywell™ Metal Hearing Protection Product Code.: 11896960
Add to basket

Sperian™ Honeywell™ Metal Hearing Protection

Product Code. 11896960
Black (Headband), Dark Gray (Cup)
Click to view available options
Color:
Black
Black (Headband), Dark Gray (Cup)
This item is not returnable. View return policy
You may only order up to the available stock. Where an alternative item has been identified it will be shown below

Product Code. 11896960

Brand: Sperian™ 1010923

Edge
Add to basket
Add to basket
This item is not returnable. View return policy
You may only order up to the available stock. Where an alternative item has been identified it will be shown below

Honeywell™ Metal noise monitoring headsets reduce background noises to safe levels whilst efficiently collecting noise exposure, helping identify risks and take action.

  • Designed to fit and seal to the workers ear perfectly
  • Reduces background noises to safe levels
  • Efficiently collect noise exposure, helping identify risks and take action
TRUSTED_SUSTAINABILITY

Specifications

Color Black (Headband), Dark Gray (Cup)
Headband Material Steel
For Use With (Application) To block noise
Product Type Hearing Protection
Headband Color Black
Material Metal
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Sperian™ Honeywell™ Metal Hearing Protection >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.