Learn More
Sperian Protection™ Honeywell™ Optifit Series Silicone Full Face Mask
Full face mask designed for customized respiratory protection depending on the canister type assembled to it. Sperian Protection™ Honeywell™ Optifit Series Silicone Full Face Mask features a silicone skirt for comfort and resistance.
Brand: Sperian Protection™ 1715021
This item is not returnable.
View return policy
Description
- Comfortable U-shaped silicone skirt
- Optimal lateral and downward vision
- Choice of filter
- APR class 2 following EN 136:1998
Specifications
EC Category PPE 3, Quality Assurance ISO 9001 / 2000, EC Certificate 0070-003-079-01-02-0009 - Ext. 01-10-03, EC Attestation Number 0070-003-079-01-02-0009 - Ext. 01-10-03, EN 136:1998 | |
Full Face Mask | |
For Use with EN141 or EN143 Class 1 or 2 Certified Sperian Filters | |
Full Lens |
Silicone | |
Large | |
RD40 connection | |
1 Ea. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Sperian Protection™ Honeywell™ Optifit Series Silicone Full Face Mask > Lens Type: Full Lens
Spot an opportunity for improvement?Share a Content Correction