missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HP1 alpha Antibody (RAB-C133), Alexa Fluor™ 594 - Chimeric, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-20127AF594
This item is not returnable.
View return policy
Description
HP1 alpha Monoclonal antibody specifically detects HP1 alpha in Human samples. It is validated for ELISA
Specifications
| HP1 alpha | |
| Monoclonal | |
| Alexa Fluor 594 | |
| Antigen p25, chromobox homolog 5, chromobox homolog 5 (Drosophila HP1 alpha), chromobox homolog 5 (HP1 alpha homolog, Drosophila), chromobox protein homolog 5, Heterochromatin protein 1 homolog alpha, heterochromatin protein 1-alpha, HP1, HP1 alpha, HP1 alpha homolog, HP1A, HP1-ALPHA, HP1Hs alpha, HP1Hs-alpha | |
| This antibody was obtained by recombinant antibody (rAb) phage display recognizing CBX5 protein under non-denaturing conditions. | |
| 0.1 mL | |
| Cellular Markers, Chromatin Research, DNA Repair, DNA replication Transcription Translation and Splicing | |
| 23468 | |
| Store at 4°C in the dark. | |
| IgG κ |
| ELISA | |
| RAB-C133 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction