Learn More
HSF1 Antibody (4B4), Janelia Fluor™ 646, Novus Biologicals™
Rat Monoclonal Antibody
Brand:  Novus Biologicals NBP2-42206JF646
Product Code. 18782683
  • 633.00 € / 0.10mL
Please to purchase this item. Need a web account? Register with us today!
Explore more special offers
This item is not returnable. View return policy
Description
Description
HSF1 Monoclonal antibody specifically detects HSF1 in Human, Mouse, Rat, Bovine, Guinea Pig, Hamster, Monkey, Rabbit samples. It is validated for ELISA, Immunohistochemistry, Immunoprecipitation, Gel Super Shift Assays
TRUSTED_SUSTAINABILITY
Specifications
Specifications
HSF1
Monoclonal
Janelia Fluor 646
50mM Sodium Borate
Rat
Protein G purified
RUO
Primary
Human, Mouse, Rat, Bovine, Guinea Pig
Purified
ELISA, Immunohistochemistry, Immunoprecipitation, Gel Supershift
4B4
ELISA, Immunohistochemistry, Immunoprecipitation, Gel Super Shift Assays
heat shock factor protein 1, heat shock transcription factor 1HSTF1HSF 1, HSTF 1
Purified recombinant mouse HSF1 protein, epitope mapping to amino acids 425-439
0.1 mL
Cancer, DNA Repair, Transcription Factors and Regulators
3297
Store at 4°C in the dark.
IgG1
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
HSF1 Antibody (4B4), Janelia Fluor™ 646, Novus Biologicals™ > 0.1 mL; Janelia Fluor 646