missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HSP90 alpha Monoclonal Antibody (5G5), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA517091
This item is not returnable.
View return policy
Description
MA5-17091 targets HSP90AA1 in indirect ELISA, FACS, IHC and WB applications and shows reactivity with Human, Mouse, and Non-human primate samples. The MA5-17091 immunogen is purified recombinant fragment of human HSP90AA1 expressed in E. Coli. MA5-17091 detects HSP90AA1 which has a predicted molecular weight of approximately 90kDa.
Heat shock proteins (HSP) are proteins that are expressed in response to various biological stress, including heat. HSP90 is a 90 kDa protein that is induced under stress conditions, but is also one of the most abundant cellular proteins found under non-stress conditions. HSP90 has been found to be associated with a number of other intracellular proteins, including steroid receptors, actin, tubulin, and some kinases. It has been suggested that HSP90 is necessary to maintain structural integrity of at least some steroid receptors to allow for ligand binding during receptor activation.Specifications
HSP90 alpha | |
Monoclonal | |
1 mg/mL | |
PBS with 0.05% sodium azide | |
P07900, P07901 | |
Hsp90aa1 | |
Purified recombinant fragment of human HSP90AA1 expressed in E. Coli. | |
100 μg | |
Primary | |
Human, Mouse, Non-human Primate | |
Antibody | |
IgG1 |
ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
5G5 | |
Unconjugated | |
Hsp90aa1 | |
86kDa; 89kDa; 90-kDa heat shock protein; 90-kDa heat shock protein alpha; akineto; AL024080; AL024147; EL52; epididymis luminal secretory protein 52; epididymis secretory sperm binding protein Li 65p; fb17b01; heat shock 86 kDa; heat shock 90kD protein 1, alpha; heat shock 90kD protein 1, alpha-like 4; heat shock 90kD protein, alpha-like 4; heat shock 90kDa protein 1, alpha; heat shock 90kDa protein 1, beta; Heat shock protein; heat shock protein 1, alpha; heat shock protein 86; heat shock protein 90; heat shock protein 90 alpha family class A member 1; heat shock protein 90, alpha (cytosolic), class A member 1; heat shock protein 90, alpha (cytosolic), class A member 1, tandem duplicate 1; heat shock protein 90-alpha 1; heat shock protein 90kDa alpha (cytosolic), class A member 1; heat shock protein 90kDa alpha family class A member 1; heat shock protein alpha; Heat shock protein HSP 90-alpha; Heat shock protein HSP 90-alpha 1; heat shock protein HSP90 alpha; heat shock protein hsp90a | |
Mouse | |
Protein G | |
RUO | |
15519, 3320 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |