Learn More
Novus Biologicals™ Human Adenosine Deaminase/ADA ELISA Kit (Chemiluminescence)
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture.
Brand:  Novus Biologicals™ NBP2-66434
Product Code. 18201773
  • 944.00 € / 1 set
Please to purchase this item. Need a web account? Register with us today!
Explore more special offers
Description
Description
The Human Adenosine Deaminase/ADA ELISA Kit (Chemiluminescence) measures human Adenosine Deaminase/ADA in serum, plasma and other biological fluids.
Specifications
Specifications
78.13-5000 pg/mL
This kit recognizes Human ADA in samples. No significant cross-reactivity or interference between Human ADA and analogues was observed.
Biological Fluids, Plasma, Serum
100
Human
Storage of components varies. See protocol for specific instructions.
46.88 pg/mL
ELISA Kit (Chemiluminescence)
ELISA
ADA
1 Kit
Adenosine Deaminase/ADA