Learn More
Novus Biologicals™ Human Adenosine Deaminase/ADA ELISA Kit (Chemiluminescence)
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture.
Brand: Novus Biologicals™ NBP2-66434
This item is not returnable.
View return policy
Description
The Human Adenosine Deaminase/ADA ELISA Kit (Chemiluminescence) measures human Adenosine Deaminase/ADA in serum, plasma and other biological fluids.Specifications
78.13-5000 pg/mL | |
This kit recognizes Human ADA in samples. No significant cross-reactivity or interference between Human ADA and analogues was observed. | |
Biological Fluids, Plasma, Serum | |
100 | |
Human | |
Storage of components varies. See protocol for specific instructions. |
46.88 pg/mL | |
ELISA Kit (Chemiluminescence) | |
ELISA | |
ADA | |
1 Kit | |
Adenosine Deaminase/ADA |